Supelco® PESTANAL® Organic Reference Standard, Tefluthrin
Catalog # 35548-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:79538-32-2
- Description:Tefluthrin
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00274607
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₄ClF₇O₂
- Molecular weight:418,73
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310