Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole-Desthio
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole-Desthio
Catalog # 32429-20MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole-Desthio
Catalog # 32429-20MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    120983-64-4
  • Description:
    Prothioconazole-desthio
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H361d - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    α-(1-Chlorocyclopropyl)-α-[(2-chlorophenyl)methyl]-1H-1,2,4-triazole-1-ethanol
  • Formula:
    C₁₄H₁₅Cl₂N₃O
  • Molecular weight:
    312,19
  • Flash point:
    Not applicable

Specifications

About this item

Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405