Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole
  34232-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Prothioconazole
  34232-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    178928-70-6
  • Description:
    Prothioconazole
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione
  • Formula:
    C₁₄H₁₅Cl₂N₃OS
  • Molecular weight:
    344,26
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS09
 : P273 - P391 - P501