Supelco® Organic Reference Standard, PCB No 209 solution
Catalogus # 41612-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:2051-24-3
- Description:PCB No 209 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055243
- M-Clarity quality level:MQ100
- Synonym(s):Decachlorobiphenyl solution
- Formula:C₁₂Cl₁₀
- Molecular weight:498,66
- Storage temperature:2...8 °C
- Flash point:–4 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331