Supelco® PESTANAL® Organic Reference Standard, Oxadiazon
Catalog # 33382-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:19666-30-9
- Description:Oxadiazon
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD00128056
- M-Clarity quality level:MQ100
- Formula:C₁₅H₁₈Cl₂N₂O₃
- Molecular weight:345,22
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS09
Caution: P273 - P391 - P501