Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Monoquat Iodide-(Methyl-D₃)
  34565-25MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Monoquat Iodide-(Methyl-D₃)
  34565-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    22884-38-4
  • Description:
    Monoquat iodide-(methyl-d₃)
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Methyl-d₃-4,4′-bipyridinium iodide; 1-Methyl-d₃-4-(4-pyridyl)pyridinium iodide; Monoquat-d3 iodide
  • Formula:
    C₁₁D₃H₈IN₂
  • Molecular weight:
    301,14
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07
 : P302 + P352 - P305 + P351 + P338