Supelco® PESTANAL® Organic Reference Standard, Methasulfocarb
Catalog # 32148-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:66952-49-6
- Description:Methasulfocarb
- Storage conditions:2...30 °C
- Hazard class description:H301 - H411
- Shipping conditions:STD
- MDL No.:MFCD00128926
- M-Clarity quality level:MQ100
- Synonym(s):S-4-(Mesyloxy)phenyl methyl(thiocarbamate); S-[4-[(Methylsulfonyl)oxy]phenyl] N-methylcarbamothioate
- Formula:C₉H₁₁NO₄S₂
- Molecular weight:261,32
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P273 - P301 + P310 + P330