Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Hexaconazol
  34348-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Hexaconazol
  34348-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    79983-71-4
  • Description:
    Hexaconazol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H317 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144441
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (±)-2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)-2-hexanol
  • Formula:
    C₁₄H₁₇Cl₂N₃O
  • Molecular weight:
    314,21
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352