Supelco® PESTANAL® Organic Reference Standard, Heptenophos
41373-25MG
: Merck
:
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:23560-59-0
- Description:Heptenophos
- Storage conditions:–10...–25 °C
- Hazard class description:H301 - H312 - H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055303
- M-Clarity quality level:MQ100
- Synonym(s):7-Chlorobicyclo[3.2.0]hepta-2,6-dien-6-yl dimethyl phosphate
- Formula:C₉H₁₂ClO₄P
- Molecular weight:250,62
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: Bottomless glass bottles are inserted with fused cone.
: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310