Supelco® PESTANAL® Organic Reference Standard, Formothion solution
46424-250MG
: Merck
:
- Concentration:~80% in xylene
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:2540-82-1
- Description:Formothion solution
- Storage conditions:2…8 °C
- Hazard class description:H226 - H302 - H304 - H311 + H331 - H315 - H319 - H335 - H373 - H412
- Shipping conditions:STD
- MDL No.:MFCD00210289
- M-Clarity quality level:MQ100
- Synonym(s):S-[2-(Formylmethylamino)-2-oxoethyl] O,O-dimethyl dithiophosphate
- Molecular weight:257,27
- Flash point:25 °C - Closed cup
- GHS signal: GHS02, GHS08, GHS07
: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P331