Supelco® Organic Reference Standard, Formaldehyde O-Pentafluorophenylmethyl-Oxime
Catalog # 41558-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:86356-73-2
- Description:Formaldehyde O-pentafluorophenylmethyl-oxime
- Seq:02
- Storage conditions:2...8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00191477
- M-Clarity quality level:MQ100
- Synonym(s):Formaldehyde O-2,3,4,5,6-PFBHA-oxime
- Formula:C₈H₄F₅NO
- Molecular weight:225,12
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Packaging: Bottomless glass bottles inserted with fused cone.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338