Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fluridon
Supelco® PESTANAL® Organic Reference Standard, Fluridon
Catalog # 45511-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fluridon
Catalog # 45511-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    59756-60-4
  • Description:
    Fluridon
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H311 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078682
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₉H₁₄F₃NO
  • Molecular weight:
    329,32
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P273 - P280 - P302 + P352 + P312 - P361 + P364 - P391 - P405