Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Ethirimol
Supelco® PESTANAL® Organic Reference Standard, Ethirimol
Catalog # 45478-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Ethirimol
Catalog # 45478-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    23947-60-6
  • Description:
    Ethirimol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H312
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055519
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₁H₁₉N₃O
  • Molecular weight:
    209,29
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P280 - P302 + P352 + P312 - P362 + P364 - P501