Pesticide reference standard, Endrin aldehyde, analytical standard, Supelco®
Catalog # 442578.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:7421-93-4
- Description:Endrin aldehyde
- Formula:C₁₂H₈Cl₆O
- Storage temperature:RT
- Description:Endrin aldehyde
- Pk Info:25 mg/ampule