Pesticide reference standard, DNC-d8, VETRANAL(TM), analytical standard, Supelco®
Catalog # 34214-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1156508-87-0
- Description:DNC-d8, VETRANAL(TM)
- M-Clarity quality level:MQ100
- Formula:C₁₃D₈H₂N₄O₅
- Storage temperature:2...8 °C
- Description:DNC-d8, VETRANAL(TM)