Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Diuron-D₆
Supelco® PESTANAL® Organic Reference Standard, Diuron-D₆
Catalog # 34018-10MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Diuron-D₆
Catalog # 34018-10MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1007536-67-5
  • Description:
    Diuron-d₆
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01632291
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-(3,4-Dichlorophenyl)-1,1-dimethylurea-d₆
  • Formula:
    C₉H₄D₆Cl₂N₂O
  • Molecular weight:
    239,13
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P264 - P270 - P273 - P301 + P312 - P308 + P313