Supelco® PESTANAL® Organic Reference Standard, Diquat Dibromide-(Ethylene-D₄)
Catalogus # 03627-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:PESTANAL®, Analytical standard
- Description:Diquat dibromide-(ethylene-d₄)
- Storage conditions:2...30 °C
- Hazard class description:H290 - H301 + H311 - H315 - H317 - H319 - H330 - H335 - H372 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):1,1′-Ethylene-d₄-2,2′-dipyridylium dibromide
- Formula:C₁₂D₄H₈Br₂N₂
- Molecular weight:348,07
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338 - P314