Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, Dihexyl phthalate
Supelco® Organic Reference Standard, Dihexyl phthalate
Catalog # 49617-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Dihexyl phthalate
Catalog # 49617-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    84-75-3
  • Description:
    Dihexyl phthalate
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H360FD
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00043720
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,2-Benzenedicarboxylic acid 1,2-dihexyl ester; Di-n-hexyl phthalate; Phthalic acid dihexyl ester
  • Molecular weight:
    334,45
  • Flash point:
    177 °C

Specifications

About this item

  • GHS signal: GHS08
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501