Supelco® PESTANAL® Organic Reference Standard, Diclobutrazol
36764-50MG
: Merck
:
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:75736-33-3
- Description:Diclobutrazol
- Storage conditions:2...30 °C
- Hazard class description:H319 - H411
- Shipping conditions:STD
- MDL No.:MFCD00144110
- M-Clarity quality level:MQ100
- Formula:C₁₅H₁₉Cl₂N₃O
- Molecular weight:328,24
- Flash point:Not applicable
- GHS signal: GHS07, GHS09
: P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P391