Supelco® PESTANAL® Organic Reference Standard, Cymoxanil
Catalog # 34326-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:57966-95-7
- Description:Cymoxanil
- Storage conditions:2...30 °C
- Hazard class description:H302 - H317 - H361fd - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00137381
- M-Clarity quality level:MQ100
- Synonym(s):1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea
- Formula:C₇H₁₀N₄O₃
- Molecular weight:198,18
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P273 - P280 - P301 + P312 - P302 + P352 - P308 + P313