Supelco® PESTANAL® Organic Reference Standard, Cycloxidim
Catalog # 31596-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:101205-02-1
- Description:Cycloxidim
- Storage conditions:–20 °C
- Hazard class description:H361d
- Shipping conditions:STD
- MDL No.:MFCD01741622
- M-Clarity quality level:MQ100
- Formula:C₁₇H₂₇NO₃S
- Molecular weight:325,47
- Storage temperature:−20 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501