Organic reference standard, Chloroform, analytical standard, Supelco®
Catalog # 48520-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:67-66-3
- Description:Chloroform
- M-Clarity quality level:MQ100
- Melting point:−63 °C (lit.)
- Formula:CHCl₃
- Description:Chloroform
- Boiling point:60.5...61.5 °C (lit.)
- Pk Info:1000 mg/ampule