Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Chlorodibromoacetic acid, analytical standard, Supelco®
Organic reference standard, Chlorodibromoacetic acid, analytical standard, Supelco®
  442519.
 :  Merck
 :  
Organic reference standard, Chlorodibromoacetic acid, analytical standard, Supelco®
  442519.
 :  Merck
 :  

 

  • Pk:
    1
  • Standards:
    Analytical standard
  • CAS No.:
    5278-95-5
  • Description:
    Chlorodibromoacetic acid
  • Melting point:
    99...102 °C (lit.)
  • Formula:
    C₂HBr₂ClO₂
  • Storage temperature:
    room temprature
  • Description:
    Chlorodibromoacetic acid
  • Pk Info:
    100 mg/ampule