Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Chlormequat-1,1,2,2-D₄ Chloride
Catalog # 00291-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Chlormequat-1,1,2,2-D₄ Chloride
Catalog # 00291-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    PESTANAL®, Analytical standard
  • Description:
    Chlormequat-1,1,2,2-d₄ chloride
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H311
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04112639
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2-Chloroethyl-d₄)trimethylammonium chloride; Chlorocholine-1,1,2,2-d₄ chloride
  • Formula:
    C₅D₄H₉Cl₂N
  • Molecular weight:
    162,09
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06
Caution: P264 - P270 - P280 - P301 + P312 - P302 + P352 + P312 - P361 + P364