Supelco® PESTANAL® Organic Reference Standard, Carbendazim-D₃
Catalog # 32413-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1255507-88-0
- Description:Carbendazim-d₃
- Storage conditions:2...30 °C
- Hazard class description:H317 - H340 - H360FD - H410
- Shipping conditions:STD
- MDL No.:MFCD17015247
- M-Clarity quality level:MQ100
- Synonym(s):Methyl-d₃ 2-benzimidazolecarbamate
- Formula:C₉D₃H₆N₃O₂
- Molecular weight:194,21
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P261 - P273 - P280 - P302 + P352 - P308 + P313