Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Butyl 4-Hydroxybenzoate-Ring-¹³C₆ Solution
  32564-1ML
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Butyl 4-Hydroxybenzoate-Ring-¹³C₆ Solution
  32564-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1416711-53-9
  • Description:
    Butyl 4-hydroxybenzoate-ring-¹³C₆ solution
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H319 - H336
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-Hydroxybenzoic acid n-butyl ester-ring-¹³C₆ solution; Butyl paraben-ring-¹³C₆ solution
  • Formula:
    ₁₃C₆C₅H₁₄O₃
  • Molecular weight:
    200,18
  • Flash point:
    –17,0 °C - closed cup

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338