Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Bromuconazol
Supelco® PESTANAL® Organic Reference Standard, Bromuconazol
Catalog # 31644-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Bromuconazol
Catalog # 31644-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    116255-48-2
  • Description:
    Bromuconazol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H361fd - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01678688
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-[[4-Bromo-2-(2,4-dichlorophenyl)tetrahydro-2-furanyl]methyl]- 1H-1,2,4-triazole
  • Formula:
    C₁₃H₁₂BrCl₂N₃O
  • Molecular weight:
    377,06
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P202 - P264 - P273 - P280 - P301 + P312 - P308 + P313