Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Bromacil
  45350-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Bromacil
  45350-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    314-40-9
  • Description:
    Bromacil
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055364
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Bromo-3-sec.-butyl-6-methyluracil
  • Formula:
    C₉H₁₃BrN₂O₂
  • Molecular weight:
    261,12
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P270 - P273 - P301 + P312 - P391 - P501