Supelco® PESTANAL® Organic Reference Standard, Brodifacoum
46036-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:56073-10-0
- Description:Brodifacoum
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H360D - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD00084803
- M-Clarity quality level:MQ100
- Molecular weight:523,42
- GHS signal: GHS08, GHS06, GHS09
: P202 - P264 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310