Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, Bis(4-Methylpentyl)Phthalate
Supelco® Organic Reference Standard, Bis(4-Methylpentyl)Phthalate
Catalog # 41260-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Bis(4-Methylpentyl)Phthalate
Catalog # 41260-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    259139-51-0
  • Description:
    Bis(4-methylpentyl)phthalate
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H360FD
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00072270
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Phthalic acid bis(4-methylpentyl) ester
  • Formula:
    C₂₀H₃₀O₄
  • Molecular weight:
    334,45
  • Storage temperature:
    2...8 °C

Specifications

About this item

  • GHS signal: GHS08
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501