Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, BDE No 205 Solution
  33688-1ML
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, BDE No 205 Solution
  33688-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    446255-56-7
  • Description:
    BDE No 205 solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD09037729
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,3,3′,4,4′,5,5′,6-Octabromodiphenyl ether solution; 2,3,3′,4,4′,5,5′,6-OctaBDE; PBDE 205
  • Formula:
    C₁₂H₂Br₈O
  • Molecular weight:
    801,38
  • Storage temperature:
    2...8 °C
  • Flash point:
    –12 °C

 

 

  • GHS signal: GHS02, GHS08, GHS07, GHS09
 : P210 - P273 - P301 + P310 + P331 - P302 + P352