Supelco® Organic Reference Standard, Arachidic Acid
Catalog # 39383-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:506-30-9
- Description:Arachidic acid
- Storage conditions:–20 °C
- Shipping conditions:DICE
- MDL No.:MFCD00002755
- M-Clarity quality level:MQ100
- Synonym(s):Arachic acid; Arachidinic acid; Eicosanoic acid; Icosanoic acid; NSC 93983
- Melting point:74...76 °C (lit.); 74...76 °C
- Formula:CH₃(CH₂)₁₈COOH
- Molecular weight:312,53
- Storage temperature:−20 °C
- Flash point:110 °C - closed cup