Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, α-Ionone
Supelco® Organic Reference Standard, α-Ionone
Catalog # 03642-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, α-Ionone
Catalog # 03642-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    127-41-3
  • Description:
    α-Ionone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H334
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001565
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one
  • Formula:
    C₁₃H₂₀O
  • Molecular weight:
    192,30
  • Density:
    0,93 g/ml at 25 °C (lit.)
  • Flash point:
    118 °C - closed cup
  • Boiling point:
    259...263 °C (lit.)

Specifications

About this item

  • GHS signal: GHS08
Caution: P261 - P342 + P311