Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Acetaldehyde Solution
Catalog # 36392-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Acetaldehyde Solution
Catalog # 36392-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    75-07-0
  • Description:
    Acetaldehyde solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H224 - H319 - H335 - H341 - H350
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00006991
  • M-Clarity quality level:
    MQ100
  • Formula:
    CH₃CHO
  • Molecular weight:
    44,05
  • Concentration:
    Natural, 50 wt, % in ethanol
  • Density:
    0,868 g/ml at 20 °C

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07
Caution: P202 - P210 - P233 - P305 + P351 + P338 - P308 + P313 - P403 + P233