Supelco® Organic Reference Standard, 4-Tert-Octylphenol-Ring-¹³C₆ Solution
Catalog # 32272-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1173020-24-0
- Description:4-tert-Octylphenol-ring-¹³C₆ solution
- Seq:02
- Storage conditions:–20 °C
- Hazard class description:H225 - H319 - H336
- Shipping conditions:WICE
- MDL No.:MFCD09842828
- M-Clarity quality level:MQ100
- Synonym(s):4-(1,1,3,3-Tetramethylbutyl)phenol-¹³C₆; 4-tert-OP-13C6
- Formula:₁₃C₆C₈H₂₂O
- Molecular weight:212,28
- Storage temperature:−20 °C
- Flash point:–17,0 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P305 + P351 + P338