Supelco® Organic Reference Standard, 4-Tert-Octylphenol-3,5-D₂ Monoethoxylate Solution
Catalog # 33525-10ML
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mL
- Standards:Analytical standard
- CAS No.:1173022-57-5
- Description:4-tert-Octylphenol-3,5-d₂ monoethoxylate solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H319 - H336
- Shipping conditions:STD
- MDL No.:MFCD09842825
- M-Clarity quality level:MQ100
- Synonym(s):2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy-3,5-d₂]ethanol; 4-tert-OP1EO-D2
- Formula:C₁₆D₂H₂₄O₂
- Molecular weight:252,39
- Storage temperature:−20 °C
- Flash point:–17 °C - Information refers to the main component,
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P261 - P280 - P304 + P340 + P312 - P370 + P378