Supelco® Organic Reference Standard, 4-Phenyl-1-Cyclohexene
Catalog # 05595-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:4994-16-5
- Description:4-Phenyl-1-cyclohexene
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00049188
- M-Clarity quality level:MQ100
- Synonym(s):3-Cyclohexen-1-ylbenzene; 4-Phenylcyclohexene
- Formula:C₁₂H₁₄
- Molecular weight:158,24
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P301 + P312 + P330