Organic reference standard, 4,4'-Diaminodiphenyl sulfide, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:139-65-1
- M-Clarity quality level:MQ100
- Melting point:105...107 °C (lit.)
- Formula:C₁₂H₁₂N₂S
- Description:4,4'-Diaminodiphenyl sulfide
Specifications
Frequently Bought Together