Supelco® Organic Reference Standard, 4-(3,5-Dimethyl-3-Heptyl)Phenol
33707-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:186825-36-5
- Description:4-(3,5-Dimethyl-3-heptyl)phenol
- Storage conditions:–20 °C
- Hazard class description:H302 - H314 - H361fd - H410
- Shipping conditions:STD
- MDL No.:MFCD11870117
- M-Clarity quality level:MQ100
- Synonym(s):353 Nonylphenol; 4-(1-Ethyl-1,3-dimethylpentyl)phenol; p-353-NP
- Formula:C₁₅H₂₄O
- Molecular weight:220,35
- Flash point:Not applicable
- GHS signal: GHS08, GHS05, GHS07, GHS09
: P201 - P273 - P280 - P301 + P330 + P331 - P303 + P361 + P353 - P305 + P351 + P338