Supelco® Organic Reference Standard, 4-(1-Ethyl-1-Methylhexyl) Phenol
32702-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:52427-13-1
- Description:4-(1-Ethyl-1-methylhexyl)phenol
- Storage conditions:–20 °C
- Hazard class description:H301 - H314 - H361fd - H410
- Shipping conditions:STD
- MDL No.:MFCD16652542
- M-Clarity quality level:MQ100
- Synonym(s):3M3-Nonylphenol isomer; 4-(3-Methyl-3-octyl)phenol
- Formula:C₁₅H₂₄O
- Molecular weight:220,35
- Storage temperature:−20 °C
- Flash point:Not applicable
- GHS signal: GHS08, GHS05, GHS07, GHS09
: P202 - P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338