Order Entry
Netherlands
ContactUsLinkComponent
Sulphuric acid 98%, LiChropur™ for HPLC, Supelco®
Sulphuric acid 98%, LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates
Sulphuric acid 98%, LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (alkalimetric):
    ≥98%
  • Identity:
    Passes test
  • Colour:
    ≤10 Hazen
  • Residue on ignition (as SO₄) :
    ≤10 ppm
  • Al (Aluminium):
    ≤0,050 ppm
  • Ca (Calcium):
    ≤0,20 ppm
  • Cu (Copper):
    ≤0,010 ppm
  • Fe (Iron):
    ≤0,10 ppm
  • K (Potassium):
    ≤0,10 ppm
  • Mg (Magnesium):
    ≤0,050 ppm
  • Na (Sodium):
    ≤0,50 ppm
  • HPLC gradient suitability test (220, 254, 280 nm) chromophoric impurities based on 4'-hydroxyacetophenone:
    ≤10 ppm
  • Chromophore purity:
    ≥99,999%

Specification Test Results

Related Information

Frequently Bought Together