Pharmaceutical analytical standard, Testosterone-16,16,17-d3 3-phenylpropionate, reference material, Honeywell Fluka™
Catalog # HONCR1538-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:876054-55-6
- Description:Testosterone-16,16,17-d₃ 3-phenylpropionate
- Formula:C₂₈H₃₃D₃O₃
- Storage temperature:Ambient
- Description:Testosterone-16,16,17-d₃ 3-phenylpropionate
- Freezing point:105...110 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results