Order Entry
Netherlands
ContactUsLinkComponent
Pharmaceutical analytical standard, Methyltestosterone-d3, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Methyltestosterone-d3, reference material, Honeywell Fluka™
Catalog # HONCR1534-10MG
CAS Number:  
Pharmaceutical analytical standard, Methyltestosterone-d3, reference material, Honeywell Fluka™
Catalog # HONCR1534-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    96425-03-5
  • Description:
    Methyltestosterone-d₃
  • Formula:
    C₂₀H₂₇D₃O₂
  • Storage temperature:
    Ambient
  • Description:
    Methyltestosterone-d₃
  • Freezing point:
    149...150 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results