Order Entry
Netherlands
ContactUsLinkComponent
Pharmaceutical analytical standard, Arprinocid, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Arprinocid, reference material, Honeywell Fluka™
Catalog # HONCR1477-10MG
CAS Number:  
Pharmaceutical analytical standard, Arprinocid, reference material, Honeywell Fluka™
Catalog # HONCR1477-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    55779-18-5
  • Description:
    Arprinocid
  • Melting point:
    246...251 °C
  • Formula:
    C₁₂H₉ClFN₅
  • Storage temperature:
    Ambient
  • Description:
    Arprinocid
  • Flash point:
    259 °C
  • Boiling point:
    504 °C
  • Freezing point:
    248 - 249 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results