Order Entry
Netherlands
ContactUsLinkComponent
 
Organic analytical standards, beta-Zearalenol-Zearalanol-d5, neat, reference material, Honeywell Fluka™
  HONCR1441-10MG
 :  
Organic analytical standards, beta-Zearalenol-Zearalanol-d5, neat, reference material, Honeywell Fluka™
  HONCR1441-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    beta-Zearalenol-Zearalanol-d₅
  • Formula:
    C₁₈H₂₁D₅O₅
  • Storage temperature:
    –20 °C
  • Description:
    beta-Zearalenol-Zearalanol-d₅
  • Freezing point:
    142...144 °C

 

 

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results