Order Entry
Netherlands
ContactUsLinkComponent
Pharmaceutical analytical standard, (±)-Ketamine-(methyl-d3) hydrochloride, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, (±)-Ketamine-(methyl-d3) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1160-10MG
CAS Number:  
Pharmaceutical analytical standard, (±)-Ketamine-(methyl-d3) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1160-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1073254-49-5
  • Description:
    (±)-Ketamine-(methyl-d₃) hydrochloride
  • Formula:
    C₁₃D₃H₁₃ClNO · HCl
  • Storage temperature:
    2...8 °C
  • Description:
    (±)-Ketamine-(methyl-d₃) hydrochloride
  • Freezing point:
    240...242 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results