VWR®, Davidson's fixative
Catalog # VWRK641139
Supplier: KLINIPATH
CAS Number:
Specifications
- Pk:5 L
- Description:Davidson's fixative
Specifications
About this item
Davidson's (sometimes called Hartmann's) fixative is a rapid fixative which gives good nuclear detail with minimal formalin pigment. Specimens cut and stain as if fixed in ordinary formalin.
- Fixation of small samples is rapid
- Contains no mercury or other metals
Davidson's fixative is particularly useful for preparing tumours, bone marrow specimens, gynecologic material, fatty breast, and medical biopsies. It may be used for testicular biopsies and for overnight fixation of fat containing lymph nodes.
Caution: Exposure to the fixative should be limited to 24 hours (tissue may be transferred to neutral buffered formalin or 70% alcohol for storage).