Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
Iodide/Iodate concentrate for dilution to 1L, 0.005 M I2 (0.01N) volumetric solution, Fluka™
  38065-1EA
 :  
 
 
Iodide/Iodate concentrate for dilution to 1L, 0.005 M I2 (0.01N) volumetric solution, Fluka™
  38065-1EA
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

 

 

  • Factor:
    0.998 - 1.002

 

 

 :
 :100 °C (1.013 hPa)
 :1.000 g/cm³ (20 °C)
 :GHS08
 :GHS07
 :Ambient
 :MFCD00011355