Order Entry
Netherlands
ContactUsLinkComponent
Rhod-2 tripotassium salt ≥90% (by HPLC) fluorescent indicator, for calcium determination
Rhod-2 tripotassium salt ≥90% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50021.
Supplier:  Biotium
CAS Number:  
Rhod-2 tripotassium salt ≥90% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50021.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :869 g/mol
Storage Temperature:Refrigerator
Formula:C₄₀H₃₉K₃N₄O₁₁