Order Entry
Netherlands
ContactUsLinkComponent
FLUO-3 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
FLUO-3 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50011.
Supplier:  Biotium
CAS Number:  
FLUO-3 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50011.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :960 g/mol
Storage Temperature:Refrigerator
CAS Number:134907-84-9
Formula:C₃₆H₂₅Cl₂K₅N₂O₁₃